Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P3817-1VL

Phospholipid Mixture for HPLC

from Soybean (Glycine max)

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$78.00

$78.00


Check Cart for Availability


Select a Size

Change View
2 ML
$78.00

About This Item

UNSPSC Code:
12352200

$78.00


Check Cart for Availability

packaging

pkg of 2 mL

concentration

in chloroform (varied)

application(s)

food and beverages

shipped in

dry ice

storage temp.

−20°C

Application

Phospholipid mixture for HPLC has been used as a reference standard for the analysis of phospholipids in subcutaneous fat from Iberian pig using high-performance liquid chromatography with evaporative light scattering detector.[1]

Analyte

Description

    L-α-Lysophosphatidylcholine from Glycine max (soybean) .3 mg/mL
    L-α-Phosphatidylcholine 1.5 mg/mL
    L-α-Phosphatidylinositol ammonium salt from Glycine max (soybean) .9 mg/mL
    L-α-Phosphatidylethanolamine from Glycine max (soybean) 1.2 mg/mL

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new high-performance liquid chromatographic method with evaporative light scattering detector for the analysis of phospholipids. Application to Iberian pig subcutaneous fat.
Narvaez-Rivas M, et al.
Journal of Chromatography A, 1218(22), 3453-3458 (2011)
Shouyun Cheng et al.
Foods (Basel, Switzerland), 8(7) (2019-07-22)
A tertiary amine (N,N-dimethylcyclohexylamine, CyNMe2) was used as a switchable hydrophilicity solvent (SHS) for extracting phospholipids (PLs) from raw cream (RC), buttermilk (BM), concentrated buttermilk (CBM), and beta-serum (BS). The SHS extractions were performed with varying solvent-sample weight ratio at
Mónica Narváez-Rivas et al.
Analytica chimica acta, 954, 140-150 (2017-01-14)
The confident identification and in-depth profiling of molecular lipid species remain to be a challenge in lipidomics analysis. In this work, an off-line two-dimensional mixed-mode and reversed-phase liquid chromatography (RPLC) method combined with high-field quadrupole orbitrap mass spectrometer (Q Exactive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service