Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N11817

Disodium methyl arsonate hexahydrate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107

grade

analytical standard

packaging

ampule of 500 mg

manufacturer/tradename

Chem Service, Inc. N-11817-500MG

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucas Schmidt et al.
Food chemistry, 255, 340-347 (2018-03-25)
A method for arsenic speciation in shark, shrimp, squid, oyster and scallop using liquid chromatography coupled to inductively coupled plasma triple quadrupole mass spectrometry (LC-ICP-MS/MS) was proposed. Suitable sensitivity and selectivity by LC-ICP-MS/MS were obtained using 10 mmol L-1 (NH4)2HPO4 diluted in
Lan Ding et al.
Toxicology and applied pharmacology, 264(1), 121-130 (2012-08-08)
Arsenic (+3 oxidation state) methyltransferase (AS3MT) is the key enzyme in the pathway for methylation of arsenicals. A common polymorphism in the AS3MT gene that replaces a threonyl residue in position 287 with a methionyl residue (AS3MT/M287T) occurs at a
Takuya Mori et al.
The Science of the total environment, 409(15), 2881-2887 (2011-05-17)
The association between oxidative stress and exposure to environmental chemicals was assessed in a group of Japanese preschool children. The concentrations of 8-hydroxy-2'-deoxyguanosine (8-OHdG), 1-hydroxypyrene (1-OHP), inorganic arsenic (iAs) and monomethylarsonic acid (MMA), and cotinine in spot urine samples, collected
José P Jesus et al.
Talanta, 85(3), 1364-1368 (2011-08-03)
An analytical procedure for multiple standard additions of arsenic species using sequential injection analysis (SIA) is proposed for their quantification in seafood extracts. SIA presented flexibility for generating multiple specie standards at the ng mL(-1) concentration level by adding different
Weihua Wen et al.
Toxicology and applied pharmacology, 254(1), 41-47 (2011-05-10)
Recent studies have shown that monomethylarsonous acid is more cytotoxic and genotoxic than arsenate and arsenite, which may attribute to the increased levels of reactive oxygen species. In this study, we used hydride generation-atomic absorption spectrometry to determine three arsenic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service