Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

843381

TSKgel® DEAE-5PW (20) Bulk Media

phase DEAE (diethylaminoethyl), diethylaminoethyl, bottle of 25 mL, 20 μm particle size

Synonym(s):

TSKgel® DEAE-5PW Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
Product 843381 is not currently sold in your country. Contact Technical Service

product line

TSKgel®

form

slurry

feature

counter ion Cl-

packaging

bottle of 25 mL

manufacturer/tradename

Tosoh 43381

parameter

20 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

DEAE (diethylaminoethyl) phase, diethylaminoethyl

particle size

20 μm

operating pH

2-13

capacity

35 g/L±10 adsorption capacity (BSA)

separation technique

anion exchange

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel DEAE-5PW media is a weak anion exchanger for biomolecule purification. Its small particle size makes it an excellent choice for polishing and high performance resolution steps. It has the same selectivity as the larger particle Toyopearl DEAE-650 resin.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

103.1 °F

flash_point_c

39.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service