Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58201C30

Supelco

SUPELCOSIL LC-Diol (5 µm) HPLC Columns

L × I.D. 25 cm × 3 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

SUPELCOSIL LC-Diol HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm

material

stainless steel column

agency

suitable for USP L20

product line

SUPELCOSIL

feature

endcapped: no

manufacturer/tradename

SUPELCOSIL

packaging

1 ea of

extent of labeling

3.5% carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 3 mm

surface area

170 m2/g

surface coverage

surface coverage 3.8 μmol/m2

matrix

silica gel, spherical particle platform
fully porous particle

matrix active group

diol phase

particle size

5 μm

pore size

120 Å pore size

pH range

2-7.5

application(s)

food and beverages

separation technique

hydrophilic interaction (HILIC)
normal phase
reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

LC-Diol columns can be used to separate proteins by gel filtration chromatography. When operated with an aqueous buffer, the diol phase can effectively shield the silica surface from interacting with proteins. A well-known use of diol columns, under normal phase conditions, is the separation of steroids and sterols.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service