Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

54235-U

Supelco

Supel-Select SAX SPE Tube

bed wt. 200 mg, volume 6 mL, pk of 30

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

material

PE frit (20 μm)
polypropylene hardware

quality

MS Friendly

composition

bed wt., 200 mg

packaging

pk of 30

technique(s)

solid phase extraction (SPE): suitable

surface area

160-420 m2/g

volume

6 mL

matrix

Quaternary amine functionalized hydrophilic modified styrene polymer particle platform

matrix active group

SAX phase

particle size

50-70 μm

pore size

80-200 Å

application(s)

food and beverages

separation technique

ion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Ion-Exchange
Sample Matrix Compatibility: Aqueous solutions (biological fluids, water)

  • Quaternary amine functionalized hydrophilic modified styrene polymer
  • Developed for the extraction of a highly broad range of compounds from aqueous samples
  • Retention predominately based on ion-exchange; however, because the phase is hydrophilic modified, it is amenable to polar compounds as well.

Application

Supel-Select SAX SPE Tube may be used as an anion exchange cartridge for the solid phase extraction of arsenic in Mexican maize tortillas analyzed by hydride generation−microwave plasma atomic emission spectrometry and high-performance liquid chromatography− inductively coupled plasma−mass spectrometry.[1]

Features and Benefits

  • Excellent sample prep performance at a lower price
  • Amenable to generic methodology - save time, money, and headache during method development
  • Greater capacity allows for smaller bed weights = smaller elution volumes = time savings in sample processing
  • Resistance to overdrying allows for more robust methdology
  • Low UV and MS extractables for lower background and greater sensitivity
  • Stringent production and QC guidelines offer greater lot-to-lot, tube-to-tube, and well-to-well consistency for improved accuracy and precision

Legal Information

Supel is a trademark of Sigma-Aldrich Co. LLC

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of total arsenic and speciation analysis in Mexican maize tortillas by hydride generation?microwave plasma atomic emission spectrometry and high-performance liquid chromatography?inductively coupled plasma?mass spectrometry
Esperanza GM, et al.
Analytical Methods : Advancing Methods and Applications, 9(13), 2059-2068 (2017)

Protocols

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service