Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52741-U

Supelco

Discovery® DSC-WCX SPE Tube

bed wt. 500 mg, volume 3 mL, pk of 54

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

material

polypropylene tube

composition

bed wt., 500 mg

packaging

pk of 54

technique(s)

solid phase extraction (SPE): suitable

surface area

480 m2/g

volume

3 mL

matrix

Silica gel base material (irregularly shaped, acid washed)

matrix active group

carboxypropyl bonding

particle size

50 μm

pore size

0.9 mL/g pore volume
70 Å pore diameter

capacity

0.15 meq/g total capacity

application(s)

food and beverages

separation technique

ion exchange

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Cation exchange
Sample Matrix Compatibility: Organic or aqueous solutions

  • A polymerically bonded, ethylenediamine triacetic acid phase with a pKa of 4.8
  • Counter ion is Na+
  • Ion exchange capacity is ∼ 0.15 meq/g
  • Carries a negative charge at pH 6.8 or above
  • A pH of 2.8 or below neutralizes this phase for easier elution of strong cationic analytes that are neutralized only at extreme basic conditions
  • Typically used when dealing with very strong cationic (high pKa) compounds that may be irreversibly retained on strong cation exchangers

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Sens. 1

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Protocols

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service