Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

52173-U

Supelco

SPE Tube Cap (encloses top of SPE tubes)

for use with 6mL SPE tube, pk of 30

Synonym(s):

PK30 CAPS FOR 6ML SPE TUBES

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.21

material

yellow polypropylene

description

Plugs top of SPE tubes

packaging

pk of 30

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

compatibility

for use with 6mL SPE tube

Looking for similar products? Visit Product Comparison Guide

Disclaimer

This product is not intended for use with centrifuge tubes.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service