Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

506885

Supelco

4-Chlorobenzoic acid

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CO2H
CAS Number:
Molecular Weight:
156.57
Beilstein/REAXYS Number:
907196
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

238-241 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

room temp

SMILES string

OC(=O)c1ccc(Cl)cc1

InChI

1S/C7H5ClO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4H,(H,9,10)

InChI key

XRHGYUZYPHTUJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

460.4 °F - closed cup

flash_point_c

238 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Erik J Rosenfeld et al.
Environmental science & technology, 41(7), 2548-2553 (2007-04-19)
A new concept is introduced to characterize and model the UV/H2O2 advanced oxidation process (AOP) in water. Similarto the Rct concept used to describe OH radical exposure per ozone dose, the ROH,UV concept is defined as the experimentally determined *OH
Xukai Li et al.
Journal of hazardous materials, 163(1), 115-120 (2008-08-01)
The aim of this research was to investigate catalytic activity of petroleum coke, activated carbon (AC) prepared from this material, Ni supported catalyst on activated carbon (Ni/AC) in the ozonation of aqueous phase p-chlorobenzoic acid (p-CBA). Activated carbon and Ni/AC
W T M Audenaert et al.
Water research, 47(7), 2387-2398 (2013-03-08)
This study experimentally examined the impact of oxidation on the properties of effluent organic matter (EfOM) using two different oxidation techniques: ozonation and UV/H2O2 treatment. Multiple surrogates for EfOM related to its spectral properties, molecular size, concentration, polarity and biodegradability
Donghai Wu et al.
Bioresource technology, 102(21), 9838-9842 (2011-09-06)
Ballast water poses a biological threat to the world's waterways by transferring aquatic species from one body of water to another. This study investigates the use of combined ultraviolet (UV)/Ag-TiO(2)+ozone (O(3)) processes for treating ballast water using Amphidinium sp. as
Haizhou Liu et al.
Environmental science & technology, 46(3), 1430-1438 (2011-12-29)
The formation of lead dioxide PbO(2), an important corrosion product in drinking water distribution systems with lead-bearing plumbing materials, has been hypothesized to involve Pb(III) intermediates, but their nature and formation mechanisms remain unexplored. This study employed the electrochemical (EC)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service