Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

506435

Supelco

ASTM® D3710 Quantitative Calibration Mix

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

agency

ASTM® D3710

packaging

pkg of 1 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Analyte

Description

    Butylbenzene 3.5 % (w/w)
    Decane 3.5 % (w/w)
    2,4-Dimethylpentane 5.8 % (w/w)
    Dodecane 3.5 % (w/w)
    Heptane 10.5 % (w/w)
    Hexane 5.8 % (w/w)
    2-Methylbutane 10.5 % (w/w)
    2-Methylpentane 5.8 % (w/w)
    Octane 5.8 % (w/w)
    Pentadecane 2.3 % (w/w)
    Pentane 8.1 % (w/w)
    Propylbenzene 4.7 % (w/w)
    Tetradecane 2.3 % (w/w)
    Toluene 11.6 % (w/w)
    Tridecane 2.3 % (w/w)
    p-Xylene 14 % (w/w)
    See All (16)

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

target_organs

Central nervous system, Central nervous system,Nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-70.0 °F

flash_point_c

-56.67 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service