Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

501247

Supelco

Argon (99.995%)

aerosol can of 4 L, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
23151816

grade

analytical standard

form

gas

packaging

aerosol can of 4 L

manufacturer/tradename

Scott Gas T104-4

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

Looking for similar products? Visit Product Comparison Guide

General description

Specifications:
Contents
: 4 liters
Pressure: 120 psig
Outlet fitting: Aerosol-type push buttom
Weight: ~100 grams
Dimensions: 2.5 x 8 inches
D.O.T. specifications: 2Q

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Suitable for qualitative analysis.

pictograms

Gas cylinder

signalword

Warning

hcodes

Hazard Classifications

Press. Gas Compr. Gas

Storage Class

2A - Gases

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical methodologies based on LC?MS/MS for monitoring selected emerging compounds in liquid and solid phases of the sewage sludg
Biox. C, et al.
MethodsX, 3, 333?342-333?342 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service