Key Documents
500771
Eppendorf® TC-50 temperature controller
AC/DC input 240 V AC
About This Item
UNSPSC Code:
41115854
Features and Benefits
Controls a CH-30 Eppendorf heater from ambient temperature to 150 °C. Temperature is set via a 10 turn potentiometer. The set point and actual temperature are displayed on an LED display. Includes remote heater on/off capability and solid state control.
Other Notes
No CE mark
Footnote
Does not have a CE mark
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)