Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

48589

Supelco

Aroclor 1248

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41121813

grade

analytical standard

packaging

ampule of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

room temp

InChI

1S/C12H6Cl4/c13-9-1-7(2-10(14)5-9)8-3-11(15)6-12(16)4-8/h1-6H

InChI key

UTMWFJSRHLYRPY-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

pictograms

Health hazardEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of rat testicular androgenesis by a polychlorinated biphenyl mixture Aroclor 1248.
Silvana A
Biology of Reproduction, 62 (6), 1882-1888 (2000)
Fast gas chromatography/mass spectrometry analysis in support of risk-based decisions.
Robbat A
Field Analytical Chemistry & Technology, 3 (1), 55-66 (1999)
Andres Martinez et al.
Environment international, 36(8), 849-854 (2009-03-10)
We report the results of the first intensive survey of polychlorinated biphenyls (PCBs) in the surficial sediment of the Indiana Harbor and Ship Canal (IHSC) in East Chicago, Indiana, a part of the Calumet River tributary of Lake Michigan that
Mercedes Fernández Santiago et al.
Toxicology letters, 163(2), 91-100 (2005-11-03)
We have demonstrated previously [Pérez-Reyes, P.L., Sánchez-Alonso, J.A., López-Aparicio, P., Recio, M.N., Pérez-Albarsanz, M.A., 2001. Different molecular capacity in the induction of apoptosis by polychlorinated biphenyl congeners in rat renal tubular cell cultures. Biosci. Rep. 6, 765-778] that the polychlorinated
Jongseol Kim et al.
Journal of microbiology and biotechnology, 18(10), 1701-1708 (2008-10-29)
Previous investigations showed that three classes of haloaromatic compounds (HACs; chlorobenzoates, chlorophenols, and chlorobenzenes) enhanced the reductive dechlorination of Aroclor 1248, judging from the overall extent of reduction in Cl atoms on the biphenyl. In the present study, we further

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service