48419
1,4-Dichlorobenzene-d4 solution
certified reference material, 2000 μg/mL in dichloromethane
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
product line
TraceCERT®
CofA
current certificate can be downloaded
packaging
ampule of 1 mL
concentration
2000 μg/mL in dichloromethane
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
single component solution
storage temp.
2-30°C
InChI
1S/C6H4Cl2/c7-5-1-2-6(8)4-3-5/h1-4H/i1D,2D,3D,4D
InChI key
OCJBOOLMMGQPQU-RHQRLBAQSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Other Notes
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Central nervous system
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service