Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442573

Supelco

Dursban

PESTANAL®, analytical standard

Synonym(s):

Chlorpyrifos, Chlorpyriphos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11Cl3NO3PS
CAS Number:
Molecular Weight:
350.59
Beilstein/REAXYS Number:
1545756
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-30°C

SMILES string

CCOP(=S)(OCC)Oc1nc(Cl)c(Cl)cc1Cl

InChI

1S/C9H11Cl3NO3PS/c1-3-14-17(18,15-4-2)16-9-7(11)5-6(10)8(12)13-9/h5H,3-4H2,1-2H3

InChI key

SBPBAQFWLVIOKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dursban is a commonly used organophosphate multipurpose pesticide[1], which can be used to control a wide range of insects in agriculture and household environment.[1] It can find applications in controlling dangerous soil pests like strawberry root weevil, click beetles and european cockchafer present in strawberry plantations.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesEnvironment

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The soil pests and their chemical and biological control on strawberry plantations in Poland
Labanowska.HB and Olszak.WR
IOBC-WPRS Bulletin 111, 26(2), 93-99 (2003)
Embryotoxicity and neurotoxicity in rats associated with prenatal exposure to Dursban
MA M, et al.
Veterinary and Human Toxicology, 34(6), 498-501 (1992)
Margaret Saunders et al.
Environmental health : a global access science source, 11 Suppl 1, S5-S5 (2012-07-13)
Organophosphate pesticides are widely used on food crops grown in the EU. While they have been banned from indoor use in the US for a decade due to adverse health effects, they are still the most prevalent pesticides in the
Daniel K Nomura et al.
Nature chemical biology, 4(6), 373-378 (2008-04-29)
Delta(9)-tetrahydrocannabinol (THC), the psychoactive ingredient of marijuana, has useful medicinal properties but also undesirable side effects. The brain receptor for THC, CB(1), is also activated by the endogenous cannabinoids anandamide and 2-arachidonylglycerol (2-AG). Augmentation of endocannabinoid signaling by blockade of
D J Clegg et al.
Journal of toxicology and environmental health. Part B, Critical reviews, 2(3), 257-279 (1999-08-03)
A panel of toxicology and medical experts was convened on 7-9 April 1997 to consider the available scientific literature on chlorpyrifos, both published and unpublished, to determine the acute and chronic toxicology reference dose (RfD). In the course of reviewing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service