Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442228

Supelco

1,2-Dichloroethane-d4

analytical standard

Synonym(s):

Ethylene-d4 dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCD2CD2Cl
CAS Number:
Molecular Weight:
102.98
MDL number:
UNSPSC Code:
41121813
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.443 (lit.)

bp

83 °C (lit.)

mp

−35 °C (lit.)

density

1.307 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

mass shift

M+4

storage temp.

room temp

SMILES string

[2H]C([2H])(Cl)C([2H])([2H])Cl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2/i1D2,2D2

InChI key

WSLDOOZREJYCGB-LNLMKGTHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

60.8 °F - closed cup - Based on data from similar materials

flash_point_c

16.0 °C - closed cup - Based on data from similar materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service