303101
Calorimetric Natural Gas Reference Standard
cylinder of 14 L, analytical standard
About This Item
Recommended Products
grade
analytical standard
form
gas
packaging
cylinder of 14 L
manufacturer/tradename
Scott Gas TNB00060-14
availability
not available in EU, Canada
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
cylinder volume
14 L
application(s)
petroleum
format
multi-component solution
Application
Features and Benefits
- Calorimetric Natural Gas Reference Standard is a multi-component gas mixture suitable for GC calibration, testing of fixed and personal gas monitors and in situ industrial stack analyzers.
- A certificate of analysis is included.
- Packaged in lightweight, non-refillable, recyclable containers.
- Portable, convenient, and easy to use.
- Manufactured gravimetrically for maximum accuracy.
- NIST traceable.
Analyte
regulator
signalword
Danger
hcodes
Hazard Classifications
Flam. Gas 1A - Press. Gas Compr. Gas
Storage Class
2A - Gases
wgk_germany
WGK 1
flash_point_f
-306.4 °F
flash_point_c
-188 °C
ppe
Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service