Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

25809

Supelco

Replacement 7 μm Stainless Steel Filter

for use with 1/4 in. gas line filter (Cat. No. 20621)

Synonym(s):

REPL. 7UM FILTER FOR TEE TYPE FILTER, EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500

General description

Use this filter to replace worn or damages filters in our gas line filter. Note that this filter will not fit into our "T"-type gas line filter.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service