Key Documents
24672
Supelco
Filter for Critical Orifices
stainless steel filter, pore size 40 μm
About This Item
UNSPSC Code:
41104008
material
bulkhead brass union
stainless steel filter
feature
(40-mesh filter)
packaging
pkg of 1 filter
Swagelok union I.D.
1/4 in.
pore size
40 μm
application(s)
environmental
industrial hygiene
compatibility
for use with critical orifice
Choose from one of the most recent versions:
Certificates of Analysis (COA)