Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22311

Supelco

Supeltex M-1 Ferrule

pkg of 10 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815

packaging

pkg of 10 ea

General description

Replace ferrules on OMI-1 tube holders periodically to reduce the risk of leaks.

  • Max. Temp.: 250 °C
  • Composition: Ceramic-filled PTFE
  • Characteristics: Ideal for connection to mass spectrometers. High reusability. Isothermal use only.

Application

Supeltex M-1 Ferrule maybe used for making connection between GC sample lines and the tube during solid adsorbent preconcentration and gas chromatographic analysis of sulfur gases.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid adsorbent preconcentration and gas chromatographic analysis of sulfur gases.
Black MS
Analytical Chemistry, 50 (7), 848-851 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service