Documents
22309
Supelco
Supeltex M-1 Packed Column Ferrule, 1/8 in. Column O.D.
I.D. 1/8 in., 1/8 in. O.D. Column, pkg of 100 ea
About This Item
UNSPSC Code:
41121815
packaging
pkg of 100 ea
I.D.
1/8 in.
fitting
1/8 in. for 1/8 in. O.D. packed column
compatibility
1/8 in. O.D. Column
General description
Characteristics: Ideal for connection to mass spectrometers. High reusability. Isothermal use only.
Material Composition: Ceramic-filled PTFE
Maximum Temperature:
250 °C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)