Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20224AST

Supelco

Astec® CYCLOBOND I 2000 SP Chiral (5 μm) HPLC Columns

L × I.D. 25 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Astec® CYCLOBOND I 2000 SP Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm

material

stainless steel column

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

0-50 °C temperature
172 bar pressure (2500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

25 cm × 4.6 mm

matrix

silica particle platform
fully porous particle

matrix active group

cyclodextrin, beta- phase

particle size

5 μm

pore size

100 Å

operating pH

3.5-7

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

On CYCLOBOND I 2000 SP, the hydroxyl groups on the surface of the β-cyclodextrin have been reacted with (S)-propylene oxide. This has the effect of extending hydrogen-bonding capabilities to accommodate analytes with chiral centers that are relatively distant from an aromatic ring structure. The (S)- form shows enhanced selectivity and efficiency for some separations, methadone, for example.

  • Bonded phase: (S)-Hydroxypropyl modified β-cyclodextrin

Resources:
CYCLOBOND FAQs
CYCLOBOND Reference Bibliography
Chiral Product Literature

Other Notes

Recommended alternative 20024AST ASTEC® CYCLOBOND 2000 Chiral HPLC Column
See technical report

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic determination of the isomeric purity of a series of dioxolane nucleoside analogues
Di Marco, M.P., et al.
Journal of Chromatography A, 645 (1), 107-114 (1993)
Chuong Pham-Huy et al.
Journal of biochemical and biophysical methods, 54(1-3), 287-299 (2003-01-25)
The enantioselective high-performance liquid chromatography (HPLC) of three racemic 3-hydroxybenzodiazepines, oxazepam (Oxa), lorazepam (Lor), and temazepam (Tem), is a difficult operation because of the spontaneous chiral inversion in polar solvent. To solve this problem, we have developed an HPLC method

Related Content

CYCLOBOND is the name given to Astec technology for bonding cyclodextrins to a high purity silica gel through a stable ether linkage.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service