Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20123AST

Supelco

Astec® CYCLOBOND I 2000 AC Chiral (5 μm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Astec® CYCLOBOND I 2000 AC Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm

material

stainless steel column

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

0-50 °C temperature
172 bar pressure (2500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

15 cm × 4.6 mm

matrix

silica particle platform

matrix active group

cyclodextrin, beta- phase

particle size

5 μm

pore size

100 Å

operating pH

3.5-7

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

CYCLOBOND I 2000 AC is used primarily for aromatic alcohols or amines that are chiral on the α or β carbon.

  • Bonded phase: Acetylated beta-cyclodextrin

Resources:
CYCLOBOND FAQs
CYCLOBOND Reference Bibliography
Chiral Product Literature

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of Beraprost sodium isomers using different cyclodextrin stationary phases
Walker, Thomas A.
Journal of Chromatography A, 633 (1-2), 97-103 (1993)
Optical resolution of flavanones by high-performance liquid chromatography on various chiral stationary phases
Krause, Martin; Galensa, Rodolf
Journal of Chromatography A, 514, 147-159 (1990)
K H Kim et al.
Archives of pharmacal research, 23(3), 230-236 (2000-07-15)
(1'R, 2R)-, (1'R, 2S)-, (1'S, 2R)- and (1'S, 2S)-alpha-hydroxymetoprolol; (2R)- and (2S)-O-desmethylmetoprolol; and (2R)- and (2S)-metoprolol acid are major metabolites of (2R)-and (2S)-metoprolol, beta-adrenergic antagonist. The focus of most chiral separation methods until now has been on determination of the
Yasith S Nanayakkara et al.
Journal of chromatography. A, 1305, 94-101 (2013-08-03)
Isochromene derivatives are very important precursors in the natural products industry. Hence the enantiomeric separations of chiral isochromenes are important in the pharmaceutical industry and for organic asymmetric synthesis. Here we report enantiomeric separations of 21 different chiral isochromene derivatives
Enantiomeric separation of functionalized ethano-bridged Troger bases using macrocyclic cyclofructan and cyclodextrin chiral selectors in high-performance liquid chromatography and capillary electrophoresis with application of principal component analysis
Weatherly, Choyce A., et al.
Journal of Chromatography. B, Biomedical Applications, 955-956, 72-80 (2014)

Related Content

CYCLOBOND is the name given to Astec technology for bonding cyclodextrins to a high purity silica gel through a stable ether linkage.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service