Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

133991

Pt-Co/Hazen/APHA Colour Reference Standard

ASTM D 1209 (ASTM colour 0)

Sign Into View Organizational & Contract Pricing


About This Item

form

liquid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

Looking for similar products? Visit Product Comparison Guide

General description

The Pt-Co/Hazen/APHA Colour Reference Standard is compliant to ASTM method D 1209.

Please note that the value stated on this page is nominal. All certified values for the current lots of all Pt-Co/Hazen/APHA Colour Reference Standards can be found on the Paragon Scientific webpage

Application

The color reference solution finds its use in routine calibration of colour measuring instruments and validation of test results.

Features and Benefits

  • ISO 9001 certified
  • Traceable to international standards
  • Available as 500 mL in tamper evident packaging
  • 12 months shelf life
  • Accompanied with a latest EU compliant MSDS available in 24 languages

replaced by

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service