Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10280-U

HayeSep® Porous Polymer Adsorbent

matrix HayeSep DB, 80-100 mesh, bottle of 75 cc

Synonym(s):

Polymer-Based Adsorbent Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
eCl@ss:
32119290
NACRES:
SB.54

form

solid

packaging

bottle of 75 cc

manufacturer/tradename

Hayes Separation Inc

parameter

290 °C temp. limit

surface area

~795 m2/g

matrix

HayeSep DB

particle size

80-100 mesh

density

~0.33 g/mL (free fall density)

Looking for similar products? Visit Product Comparison Guide

General description

HayeSep porous polymers, considered second generation materials, are consistent batch-to-batch, with minimal shrinkage and monomer bleed.

For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents
Porous polymers are the most suitable adsorbent for applications based on the analysis of gases, acids, amines, organics of low carbon number and water.[1] They are best suited for gas chromatography and are basically copolymers of polydivinylbenzene (DVB) which are very porous in nature ranging from mesoporous to microporous.[2] HayeSep® has a high surface area owing to its micropores thereby making it a suitable candidate for separation of gases and volatile organic compounds (VOCs). [3] Furthermore they are relatively inert and exhibit hydrophobicity.[4]

Application

HayeSep® DB was used for analysis of nitrogen (N2), oxygen (O2), carbon monoxide (CO), nitric oxide (NO), nitrous oxide (N2O), and water vapor (H2O).[5] HayaSep® DB in combination with HayeSep Q was also used for optimisation of a gas chromatographic method for trace gaseous impurities like dioxygen (O2), dinitrogen (N2), carbon dioxide (CO2), carbon monoxide (CO), sulfur hexafluoride (SF6), methane (CH4), nitrous oxide (N2O) and carbon tetrafluoride (CF4).[6]

Footnote

HayeSep DB is a deactivated version of HayeSep D for use with basic compounds.

Legal Information

HayeSep is a registered trademark of Hayes Separation Inc.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modern Practice of Gas Chromatography.
Grob RL and Eugene FB.
Science, 79-83 (2004)
Improvements in the Determination of Decomposition Gases from 1, 3, 3-Trinitroazetidine and 5-Nitro-2, 4-dihydro-3H-1, 2, 4-trizol-3-one Using Capillary Gas Chromatography-Mass Spectrometry.
Zheng W, et al.
Journal of Chromatographic Science, 35 (10), 478-482 (1997)
Johannes Petrus de Coning et al.
Journal of chromatography. A, 1180(1-2), 151-158 (2007-12-25)
Highly reactive fluorinated gaseous matrices require special equipment and techniques for the gas chromatographic analysis of trace impurities in these gases. The impurities that were analysed at the low mg/L levels included dioxygen (O2), dinitrogen (N2), carbon dioxide (CO2), carbon
Gas Chromatography in Air Pollution Analysis.
Berezkin V.G and Drugov Y.S.
J. Chromatogr. Library, 50-51 (1991)
Trace Analysis of Specialty and Electronic Gases.
Geiger W.M and Raynor M.W.
Science, 256-257 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service