Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W2768

Sigma-Aldrich

Anti-phospho-WAVE-1 (pSer397) antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-SCAR1, Anti-WASF1, Anti-WASP Family, Member 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Product W2768 is not currently sold in your country. Contact Technical Service

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~75 kDa

species reactivity

human (predicted), mouse, rat (predicted)

concentration

~1.5 mg/mL

technique(s)

western blot: 1-2 μg/mL using mouse brain extract (S1 fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer397)

Gene Information

human ... WASF1(8936)
mouse ... Wasf1(83767)
rat ... Wasf1(294568)

General description

WAS protein family member 1 (WAVE-1) is encoded by the gene mapped to human chromosome 6q21. The encoded protein contains small WH2 acidic domain, a cofilin-like domain, proline-rich domain at C-terminal end and EVH1 domain at N-terminal end. WAVE-1 is active mainly in the brain and lungs, but its mRNA is also found in the placenta, liver and pancreas.

Biochem/physiol Actions

WAS protein family member 1 (WAVE-1) functions as a key pro-autophagic protein. The protein has an ability to increase cell survival rate. In addition, it also controls chemoresistance in leukemia cells via Beclin1/ B-cell lymphoma 2 (Bcl)-2 and Beclin1/ phosphatidylinositol 3-kinases (PI3K)-III complex-dependent pathways. The encoded protein enhances ovarian cancer cell proliferation, migration and invasion via activating PI3K/AKT (protein kinase B) and mitogen activated protein kinase p38 (p38MAPK) signaling pathways.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Knockdown of WAVE1 enhances apoptosis of leukemia cells by downregulating autophagy.
Zhang Z
International Journal of Oncology, 48, 2647-2656 (2016)
WAVE1 gene silencing via RNA interference reduces ovarian cancer cell invasion, migration and proliferation.
Zhang J
Gynecologic Oncology, 130, 354-361 (2013)
WASF4: A new member of the WAS gene family
Klimov EA
Molekuliarnaia Biologiia, 42, 625-628 (2008)
Angel Matias Sanchez et al.
Molecular endocrinology (Baltimore, Md.), 27(4), 693-702 (2013-03-15)
Sex steroids are important regulators of neuronal cell morphology, and this is critical for gender differences in brain function and dysfunction. Neuronal morphology is controlled by multiprotein complexes including moesin (a member of the ezrin/radixin/moesin family), focal adhesion kinase (FAK)
Ivonne Denise Uzair et al.
Neuroendocrinology, 110(6), 535-551 (2019-09-12)
Synaptic plasticity is the neuronal capacity to modify the function and structure of dendritic spines (DS) in response to neuromodulators. Sex steroids, particularly 17β-estradiol (E2) and progesterone (P4), are key regulators in the control of DS formation through multiprotein complexes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service