Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W1770

Sigma-Aldrich

WR99210

Synonym(s):

1,6-Dihydro-6,6-dimethyl-1-[3-(2,4,5-trichlorophenoxy)propoxy]-1,3,5-triazine-2,4-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18Cl3N5O2
CAS Number:
Molecular Weight:
394.68
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: 0.2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC1(C)N=C(N)N=C(N)N1OCCCOc2cc(Cl)c(Cl)cc2Cl

InChI

1S/C14H18Cl3N5O2/c1-14(2)21-12(18)20-13(19)22(14)24-5-3-4-23-11-7-9(16)8(15)6-10(11)17/h6-7H,3-5H2,1-2H3,(H4,18,19,20,21)

InChI key

MJZJYWCQPMNPRM-UHFFFAOYSA-N

Biochem/physiol Actions

Potent dihydrofolate reductase (DHFR) inhibitor.
WR99210 is a potent inhibitor of Plasmodium falciparum dihydrofolate reductase (DHFR), which is a major malarial drug target. It has subnanomolar potency for the wild type, double mutant and quadruple mutant dihydrofolate reductases.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E G Hankins et al.
Molecular and biochemical parasitology, 117(1), 91-102 (2001-09-12)
We have expressed dhfr alleles of Plasmodium falciparum in the budding yeast, Saccharomyces cerevisiae, and used this yeast model to identify single amino acid substitutions that confer high level pyrimethamine resistance on the background of the triple mutant dhfr (I51+R59+N108).
T F de Koning-Ward et al.
Molecular and biochemical parasitology, 117(2), 155-160 (2001-10-19)
The limited number of selectable markers available for malaria transfection has hindered extensive manipulation of the Plasmodium falciparum genome and subsequently thorough genetic analysis of this organism. In this paper, we demonstrate that P. falciparum is highly sensitive to the
G Rastelli et al.
Bioorganic & medicinal chemistry, 8(5), 1117-1128 (2000-07-06)
The nature of the interactions between Plasmodium falciparum dihydrofolate reductase (pfDHFR) and antimalarial antifolates, i.e., pyrimethamine (Pyr), cycloguanil (Cyc) and WR99210 including some of their analogues, was investigated by molecular modeling in conjunction with the determination of the inhibition constants
Alyson Auliff et al.
The American journal of tropical medicine and hygiene, 75(4), 617-621 (2006-10-14)
The increasing use of sulfadoxine-pyrimethamine (SP) for the treatment of chloroquine-resistant Plasmodium falciparum has resulted in increased reports of SP resistance of P. falciparum worldwide. Selection of SP-resistant Plasmodium vivax in areas where P. falciparum and P. vivax co-exist is
D A Fidock et al.
Molecular pharmacology, 54(6), 1140-1147 (1998-12-18)
The lack of suitable antimalarial agents to replace chloroquine and pyrimethamine/sulfadoxine threatens efforts to control the spread of drug-resistant strains of the malaria parasite Plasmodium falciparum. Here we describe a transformation system, involving WR99210 selection of parasites transformed with either

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service