Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

W0392

Sigma-Aldrich

Anti-WAVE antibody produced in rabbit

~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-WASF1, Anti-Wiskott-Aldrich syndrome protein family member 1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$609.00

$609.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$609.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$609.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 75 kDa

species reactivity

rat, human, mouse

concentration

~1.5 mg/mL

technique(s)

microarray: suitable
western blot: 0.2-0.4 μg/mL using rat and mouse brain cytosolic fraction (S1)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WASF1(8936)
mouse ... Wasf1(83767)
rat ... Wasf1(294568)

Related Categories

General description

The WASP family members include WASP, N-WASP and WAVE (WASP-family verprolin homologous protein) proteins. WAVE proteins (also termed SCAR, WASF proteins), include three isoforms WAVE-1, WAVE-2 and WAVE-3. Expression of WAVE-1 and WAVE-3 isoforms is restricted to the brain, while WAVE-2 is ubiquitously expressed. WAVE-1-3 have structures like WASP and N-WASP; these include two conserved domains: a WAVE homology domain (WHD), also known as SCAR homology domain (SHD), a C-terminal acidic domain that binds to the Arp2/3 complex; and a central core domain composed of proline-rich sequences that interact with cofilin and various SH3 proteins.

Immunogen

synthetic peptide corresponding to amino acids 540-559 located at the C-terminus of human WAVE-1. The sequence is identical in several species, including mouse and rat WAVE-1 and is conserved (75% sequence identity) in other WAVE isoforms.

Application

Anti-WAVE antibody produced in rabbit has been used in immunoblotting.

Biochem/physiol Actions

WAVE proteins also play key roles in the induction of various actin remodelling processes including membrane ruffling and lamellipodia formation. WAVE proteins bind to phosphatidylinositol-3,4,5 triphosphate (PI-3,4,5P3) formed by PI3-kinase, thus facilitating WAVE translocation to the plasma membrane. WAVE proteins are thought to mediate Rac activity indirectly via the target protein IRSp53 or by binding to a macromolecular complex that includes PIR121, Nck-associated protein 1 (Nap1), Abi, and HSPC300 proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IRSp53 is an essential intermediate between Rac and WAVE in the regulation of membrane ruffling
Miki H, et al.
Nature, 408(6813), 732-732 (2000)
Purification and architecture of the ubiquitous Wave complex
Gautreau A, et al.
Proceedings of the National Academy of Sciences of the USA, 101(13), 4379-4383 (2004)
Mechanism of regulation of WAVE1-induced actin nucleation by Rac1 and Nck
Eden S, et al.
Nature, 418(6899), 790-790 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service