W0267
Anti-WAVE-1 antibody produced in rabbit
~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-WASF1, Anti-Wiskott-Aldrich syndrome protein family member 1
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 75 kDa
species reactivity
mouse, human, rat
concentration
~2 mg/mL
technique(s)
indirect immunofluorescence: suitable
western blot: 0.25-0.5 μg/mL using rat and mouse brain cytosolic fraction (S1)
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... WASF1(8936)
mouse ... Wasf1(83767)
rat ... Wasf1(294568)
Immunogen
Application
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service