Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TGO

Sigma-Aldrich

ThermaGo

increases PCR specificity, inhibits non-specific polymerase activity, prevents mis-priming during PCR

Synonym(s):

Multiplex PCR, PCR Additive, hot start DNA polymerase, hot start PCR, hotstart

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

lyophilized powder

packaging

units of 2500 units
units of 500 units
pkg of One unit of ThermaGo is defined as the amount required for maximum improvements in amplification reactions containing 1 unit of Taq DNA polymerase in a volume of 25 μl. 

technique(s)

PCR: suitable

color

opaque

shipped in

ambient

storage temp.

2-8°C

General description

ThermaGo is a double-stranded, modified nucleic acid that terminates the mispriming by interacting with DNA polymerase during the annealing and extension steps of PCR. It improves the specificity of Taq DNA polymerase. ThermaStop and ThermaGo act together during PCR amplification at different temperatures to increase specificity and prevent errors.

Application

ThermaGo is suitable for:
  • monoplex and multiplexed end-point single nucleotide polymorphism (SNP) genotyping
  • detection of low-copy target numbers during single-cell PCR and digital PCR
  • detection of low-copy target numbers in liquid biopsies/tumor samples
  • enhancing the accuracy of multiplex PCR for next-generation sequencing and other post PCR applications

Features and Benefits

  • Acts at higher temperatures during annealing and elongation steps
  • Improves the reproducibility
  • Detects low-copy number targets by suppressing non-specific targets
  • Minimizes signal scatter in digital & standard PCR
  • Minimizes mispriming and chimera production before next-generation sequencing (NSG)
  • Enhances end-point genotyping

Reconstitution

Dissolve in Tris pH 8.3 buffer

Legal Information

ThermaGo is a trademark of ThermaGenix Inc.
ThermaStop is a trademark of ThermaGenix Inc.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

PCR additive added to master mix that stops nonspecific bands after cool down.

How to stop nonspecific bands and off target primers in PCR.

PCR additive interacts with reverse transcriptase at low temperatures to reduce priming problems that lead to nonspecific bands.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service