Key Documents
T3531
Microcentrifuge tubes, extended capacity
volume 2.0 mL , graduated, siliconized polypropylene
About This Item
UNSPSC Code:
41121703
eCl@ss:
32119210
material
cap
siliconized polypropylene
packaging
pack of 200
volume
2.0 mL , graduated
General description
The tubes listed below are specifically designed to hold more volume than other comparable tubes while still fitting all of the most commonly used microcentrifuges. Writing surface on flat lids and side wall for quick sample ID.
The lids provide an easily punctured syringe port for easy access to the contents without opening the tube. Siliconized tubes simplify research requiring low adhesion surface.
Choose from one of the most recent versions:
Certificates of Analysis (COA)