Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

T3154

Sigma-Aldrich

Tris-Phosphate-EDTA Buffer 10X

suitable for gel electrophoresis (after dilution to working concentration)

Synonym(s):

TPE Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

sterility

0.2 μm filtered

suitability

suitable for gel electrophoresis (after dilution to working concentration)

foreign activity

DNase, RNase, none detected

Application

Tris-Phosphate-EDTA Buffer 10X is diluted to working concentration and used for gel electrophoresis and gene expression studies.[1]

Other Notes

0.8 M Tris phosphate, pH approx. 8.0, 0.02 M EDTA;

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sambrook, J., et al.
Molecular Cloning: A Laboratory Manual, 6-6 (1989)
Philip Roelandt et al.
Nature protocols, 5(7), 1324-1336 (2010-07-03)
Differentiation of stem cells to hepatocytes has industrial applications, as well as the potential to develop new therapeutic strategies for liver disease. The protocol described here, sequentially using cytokines that are known to have a role in liver embryonic development

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service