SRP6157
BMP-7 human
recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)
Synonym(s):
Bone morphogenetic protein 7, Eptotermin alfa, OP1, Osteogenic protein 1
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
General description
Bone morphogenetic protein-7 (BMP-7) belongs to the transforming growth factor-β (TGF-β) superfamily. The gene encoding it is localized on human chromosome 20q13.
Biochem/physiol Actions
As implied by their name, bone morphogenetic proteins (BMPs) promote and regulate bone development, growth, remodeling and repair, in both prenatal development and postnatal growth of eye, heart, kidney, skin, and other tissues. Based on its expression early in embryogenesis, BMP-7 has a proposed role in early development. In addition, the fact that BMP-7 is closely related to BMP5 has led to speculation of possible bone inductive activity. BMP-7 induces apoptosis in B-cells. It has a role in organ development and may have a role in liver regeneration. Mutations in the gene encoding the protein have been shown to affect the development of teeth, palate and other orofacial complexes.
Physical form
Lyophilized from a solution of 50 mM sodium acetate, pH 4.5, with 50 mM NaCl.
Preparation Note
Centrifuge the vial prior to opening.
Reconstitution
Reconstitute in sterile PBS containing 0.1% endotoxin-free, recombinant human serum albumin.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
TGF-beta superfamily members and ovarian follicle development.
Knight PG and Glister C
Reproduction (Cambridge, England), 132(2), 191-206 (2006)
Yan Wang et al.
Experimental and molecular pathology, 98(3), 393-402 (2015-03-17)
This study presented the effect of bone morphogenic protein-7 (BMP-7) inhibiting epithelial-mesenchymal transition (EMT) in silicosis model. In vivo, Wistar rats were exposed to silica by intratracheal instillation. Seven days later rats were treated with BMP-7. Rats were sacrificed at
Applications of small molecule BMP inhibitors in physiology and disease.
Hong CC and Yu PB
Cytokine & Growth Factor Reviews, 20(5-6), 409-418 (2009)
Endre Kristóf et al.
Scientific reports, 5, 12540-12540 (2015-07-28)
Laser-scanning cytometry is presented as a tool allowing population scale analysis of ex vivo human brown adipogenic differentiation. It combines texture analysis and detection of Ucp1 protein content in single brown adipocytes of mixed cell populations with gene expression pattern
BMP7 expression correlates with secondary drug resistance in mantle cell lymphoma.
Camara-Clayette V
PLoS ONE, 8(9), e73993-e73993 (2013)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service