Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP6068

Sigma-Aldrich

Cyclophilin H human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE)

Synonym(s):

CYP20, CYPH, PPIH, Peptidyl prolyl isomerase H, Rotamase H

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

assay

≥95% (SDS-PAGE)

form

liquid

mol wt

19.2 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PPIH(10465)

General description

Cyclophilin H is a member of peptidyl-propyl cis-trans isomerase (PPIase) family, which catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerates the folding of proteins. The cyclophilin H is a specific component of the human U4/U6 small nuclear ribonucleoprotein particle involved in the nuclear splicing of pre-mRNA. It stably associates with the U4/U6-60kD and -90kD proteins, the human orthologues of the Saccharomyces cerevisiae Prp4 and Prp3 splicing factors.

Physical form

1 mg/mL solution in PBS (pH 7.4) containing 10% glycerol.

Preparation Note

Centrifuge the vial prior to opening.

Other Notes

MAVANSSPVN PVVFFDVSIG GQEVGRMKIE LFADVVPKTA ENFRQFCTGE FRKDGVPIGY KGSTFHRVIK DFMIQGGDFV NGDGTGVASI YRGPFADENF KLRHSAPGLL SMANSGPSTN GCQFFITCSK CDWLDGKHVV FGKIIDGLLV MRKIENVPTG PNNKPKLPVV ISQCGEM

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service