Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP5017

Sigma-Aldrich

CK2 α 2, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CK2A2, CKII-α 2, CSNK2A2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

63-72 nmol/min·mg

mol wt

~64 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CSNK2A2(1459)

General description

CK2α2 or casein kinase II alpha 2 is a member of the CK2 family of Ser/Thr protein kinases. CK2a2 plays a fundamental role in cell function and is involved in DNA replication, regulation of basal and inducible transcription, translation and control of metabolism. CK2α2 prefers utilization of acidic proteins such as caseins as substrates. The CK2a2 holoenzyme is a tetramer composed of an alpha chain, an alpha′ and two beta chains. The alpha and alpha′ chains contain the catalytic site. CK2α2 is also a component of CK2-SPT16-SSRP1 complex comprised of SSRP1, SUPT16H, CSNK2A1, CSNK2A2 and CSNK2B. This complex associates following UV irradiation. CK2a2 act as a candidate gene for inherited abnormalities of sperm morphogenesis.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Xu et al.
Nature genetics, 23(1), 118-121 (1999-09-02)
Protein kinase casein kinase II (Ck2) is a cyclic-AMP and calcium-independent serine-threonine kinase that is composed of two catalytic subunits (alpha and alpha') and two regulatory beta-subunits. Ck2 is not a casein kinase in vivo, but over 100 substrates are
D M Keller et al.
Molecular cell, 7(2), 283-292 (2001-03-10)
Phosphorylation of the human p53 protein at Ser-392 has been shown to be responsive to UV but not gamma irradiation. Here we describe identification and purification of a mammalian UV-activated protein kinase complex that phosphorylates Ser-392 of p53 in vitro.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service