Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP4029

Sigma-Aldrich

Eotaxin from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

CCL11, Eosinophil chemotactic protein chemokine (C-C motif) ligand 11, SCYA11, Small inducible cytokine A11

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

biological source

mouse

recombinant

expressed in E. coli

assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

~8.4 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

Gene Information

mouse ... Ccl11(20292)

General description

Eotaxin is a member of the CC chemokine family, and belongs to the MCP/eotaxin subfamily. It has a wide range of tissue expression, such as thymus, heart, lungs, lymph nodes, kidney, and intestines. It is also expressed by epithelial cells of lung, pleural mesothelial cells, epithelial cells of bronchial airway, and smooth muscle cells (SMCs).[1]

Biochem/physiol Actions

Eotaxin is a chemoattractant for eosinophils, and is involved inflammatory diseases, such as asthma. It is induced by glucocorticoids and cytokines.[2] Along with interleukin-5 (IL-5) it results in airway hyperresponsiveness (AHR), and hence, lung inflammatory disorders. In humans, it leads to respiratory syncytial virus (RSV) vaccine-enhanced disease, and the eosinophils are also responsible for the elimination of virus.[3] It might also be involved in the pathophysiology of inflammatory bowel diseases which involve eosinophils.[4] It can lead to the production of reactive oxygen species, thus, resulting in damaged tissue.[5] It is overexpressed in multiple diseases such as allergic rhinitis, nasal polyposis, endometriosis and Hodgkin′s disease.[1]

Physical form

Sterile filtered and then lyophilized with no additives. The protein may appear as a haze or film that is difficult to see at the bottom of the vial.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1 μg/μL. It is recommended that further dilutions be made into buffer containing carrier protein or medium containing serum.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E P Bowman et al.
The Journal of experimental medicine, 191(8), 1303-1318 (2000-04-19)
Developing B cells undergo dramatic changes in their responses to chemoattractant cytokines (chemokines) and in expression of chemokine receptors. Bone marrow pre-pro-B cells (AA4.1(+)/natural killer 1.1(-) Fraction A cells) and cells capable of generating pro-B colonies in the presence of
Xiang Zhu et al.
BMC immunology, 14, 24-24 (2013-05-28)
Proton currents are required for optimal respiratory burst in phagocytes. Recently, HVCN1 was identified as the molecule required for the voltage-gated proton channel activity associated with the respiratory burst in neutrophils. Although there are similarities between eosinophils and neutrophils regarding
J Elsner et al.
European journal of immunology, 26(8), 1919-1925 (1996-08-01)
Increased numbers of eosinophils are found in parasitic infections, autoimmune diseases and allergic diseases such as allergic asthma. They are activated by distinct cytokines and chemokines leading to the immigration in the inflamed tissue and mediate tissue damage by releasing
M Kitaura et al.
The Journal of biological chemistry, 271(13), 7725-7730 (1996-03-29)
The CC chemokine eotaxin is a selective chemoattractant for guinea pig eosinophils, first purified from bronchoalveolar lavage fluid in a guinea pig model of allergic airway inflammation. We have now isolated the gene and cDNA for a human counterpart of
E A Garcia-Zepeda et al.
Genomics, 41(3), 471-476 (1997-05-01)
Eotaxin is a CC chemokine that is a specific chemoattractant for eosinophils and is implicated in the pathogenesis of eosinophilic inflammatory diseases, such as asthma. We describe the genomic organization, complete sequence, including 1354 bp 5' of the RNA initiation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service