SRP2103
Arginine Splicing Factor, His tagged human
recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Synonym(s):
ASF, FLJ53078, MGC5228, SF2, SF2p33, SFRS1, SRp30a
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.26
Recommended Products
biological source
human
recombinant
expressed in E. coli
assay
≥80% (SDS-PAGE)
form
frozen liquid
mol wt
~29.3 kDa
packaging
pkg of 10 μg
storage condition
avoid repeated freeze/thaw cycles
concentration
650 μg/mL
color
clear
colorless
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... SRSF1(6426)
Biochem/physiol Actions
ASF or called SF2, a member of the SR protein family, is an essential pre-mRNA splicing factor required for both single and alternative splicing. Phosphorylation on serine residues located within the SR domain directly regulates ASF/SF2 activity and compartmentalization of other SR splicing factors. In addition to interacting with RNA and other splicing factors, such as U1-70K, U2AF and other SR proteins, ASF/SF2 also directly or indirectly interacts with HIV regulatory protein Rev, the C-terminal domain (CTD) of the largest subunit of RNA polymerase II, and numerous transcription factors, thereby suggesting a potential role of ASF/SF2 in coordinating of transcription and pre-mRNA splicing.
Physical form
Clear and colorless frozen liquid solution
Preparation Note
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
H Ge et al.
Cell, 62(1), 25-34 (1990-07-13)
SV40 early pre-mRNA is alternatively spliced by utilization of two different 5' splice sites and a shared 3' splice site to produce large T and small t mRNAs. The ratio of small t to large T mRNAs produced in human
A R Krainer et al.
Cell, 66(2), 383-394 (1991-07-26)
SF2 is a protein factor essential for constitutive pre-mRNA splicing in HeLa cell extracts and also activates proximal alternative 5' splice sites in a concentration-dependent manner. This latter property suggests a role for SF2 in preventing exon skipping, ensuring the
A M Zahler et al.
Genes & development, 6(5), 837-847 (1992-05-01)
We demonstrate that four different proteins from calf thymus are able to restore splicing in the same splicing-deficient extract using several different pre-mRNA substrates. These proteins are members of a conserved family of proteins recognized by a monoclonal antibody that
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service