Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0703

Sigma-Aldrich

LYS6K2

recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$527.00

$527.00


Usually ships in 2 weeks.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
25 MG
$527.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$527.00


Usually ships in 2 weeks.

Notify Me

Get notified when this item is ready to ship via email.

assay

>95% (HPLC)

form

lyophilized powder

mol wt

~445.4

shipped in

ambient

storage temp.

−20°C

Biochem/physiol Actions

A selective inhibitor of the p70 ribosomal S6 kinase (S6K). Blocks the phosphorylation of S6 at concentrations as low as 0.1-0.3 uM with no activity at GSK3beta and Erk1/2 at 10 uM (2). Useful tool to dissect insulin receptor signaling pathways and was employed to show that the action of mTORC1 on SREB P-1c expression is not mediated by S6K. Cell permeable.

Reconstitution

Formulated as a lyophilized powder. May be dissolved in DMSO (1 mg/mL) or DMF (3 mg/mL, warm).

Storage and Stability

Store desiccated as supplied at -20°C for up to 3 years. Store solutions at -20°C for up to 3 months.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service