Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0480

Sigma-Aldrich

WDR9 (1308-1436) human

recombinant, expressed in E. coli, ≥82% (SDS-PAGE)

Synonym(s):

BRWD1, N143, bromodomain and WD repeat domain containing 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$761.00

$761.00


Usually ships in 2 weeks.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$761.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$761.00


Usually ships in 2 weeks.

Notify Me

Get notified when this item is ready to ship via email.

biological source

human

recombinant

expressed in E. coli

assay

≥82% (SDS-PAGE)

form

aqueous solution

mol wt

16.2 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BRWD1(54014)

General description

Human bromodomain and WD repeat domain containing 1 (BRWD1) or WDR9 (GenBank Accession No. NM_018963) amino-acids 1308 - 1436 with N-terminal HIS-tag, MW = 16.2 kDa, expressed in an E. coli expression system.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Veronica C Ramos et al.
Biochimica et biophysica acta, 1577(3), 377-383 (2002-10-03)
We report the isolation and characterisation of the gene WDR9 (WD Repeat 9), located in the Down Syndrome critical region-2 (DCR-2) from the human chromosome 21. This gene spans 125 kb of genomic sequence and is organised in 41 exons
He Huang et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 227(4), 608-614 (2003-07-31)
Human WDR9 has been mapped to chromosome 21, within one of the Down syndrome (DS) critical regions. Here, we study the expression pattern of the murine Wdr9 gene and its protein product. We show that Wdr9 is broadly expressed in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service