Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0365

Sigma-Aldrich

p70S6K active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)

Synonym(s):

PS6K, RPS6KB1, p70(S6K)-alpha, ribosomal protein S6 kinase, 70kDa, polypeptide 1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$563.00

$563.00


Usually ships in 2 weeks.

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
20 μG
$563.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$563.00


Usually ships in 2 weeks.

Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

74 kDa

packaging

pkg of 20 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RPS6KB1(6198)

General description

Human p70S6K (GenBank Accession No. NM_003161), amino acid 1-421 with N-terminal GST tag, MW=74 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salvatore Di Paolo et al.
American journal of kidney diseases : the official journal of the National Kidney Foundation, 49(3), 462-470 (2007-03-06)
The clinical challenge for the application of rapamycin and its derivatives as anticancer drugs is the ability to prospectively identify which tumors will be sensitive to mammalian target of rapamycin (mTOR) inhibition. The present study is designed to explore mTOR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service