SRP0175
NSD2 catalytic domain Active human
recombinant, expressed in E. coli, ≥70% (SDS-PAGE)
Synonym(s):
MMSET, Multiple myeloma SET domain-containing protein, Nuclear SET domain-containing protein 2, REIIBP, Wolf-Hirschhorn syndrome candidate 1
About This Item
biological source
human
recombinant
expressed in E. coli
assay
≥70% (SDS-PAGE)
form
aqueous solution
mol wt
62 kDa
packaging
pkg of 50 μg
concentration
>0.02 mg/mL
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... WHSC1(7468)
General description
Human NSD2 (MMSET) (GenBank Accession No. NM_133330), amino acids 941-1240 with N-terminal GST tag, MW = 62kDa, expressed in Escherichia coli.
Application
Biochem/physiol Actions
Physical form
Preparation Note
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service