MilliporeSigma
  • Differential effect of MMSET mRNA levels on survival to first-line FOLFOX and second-line docetaxel in gastric cancer.
Differential effect of MMSET mRNA levels on survival to first-line FOLFOX and second-line docetaxel in gastric cancer.
British journal of cancer (2014-05-09)
J Wei, C Costa, J Shen, L Yu, J J Sanchez, X Qian, X Sun, Z Zou, A Gimenez-Capitan, G Yue, W Guan, R Rosell, B Liu
ABSTRACT
Breast cancer susceptibility gene 1 (BRCA1) expression differentially affects outcome to platinum- and taxane-based chemotherapy. Mediator of DNA damage checkpoint protein 1 (MDC1), p53-binding protein 1 (53BP1), multiple myeloma SET domain (MMSET) and ubiquitin-conjugating enzyme 9 (UBC9) are involved in DNA repair and could modify the BRCA1 predictive model. Mediator of DNA damage checkpoint protein 1, 53BP1, MMSET and UBC9 mRNA were assessed in gastric tumours from patients in whom BRCA1 levels had previously been determined. In vitro chemosensitivity assay, MMSET levels were higher in docetaxel-sensitive samples. In a separate cohort, survival was longer in those with low MMSET (12.3 vs 8.8 months; P=0.04) or UBC9 (12.4 vs 8.8 months; P=0.01) in patients receiving only folinic acid, fluorouracil (5-FU) and oxaliplatin (FOLFOX). Conversely, among patients receiving second-line docetaxel, longer survival was associated with high MMSET (19.1 vs 13.9 months; P=0.003). Patients with high MMSET and BRCA1 attained a median survival of 36.6 months, compared with 13.9 months for those with high BRCA1 and low MMSET (P=0.003). In the multivariate analyses, low MMSET (hazard ratio (HR), 0.59; P=0.04) and low UBC9 (HR, 0.52; P=0.01) levels were markers of longer survival to first-line FOLFOX, whereas palliative surgery (HR, 2.47; P=0.005), low BRCA1 (HR, 3.17; P=0.001) and low MMSET (HR, 2.52; P=0.004) levels were markers of shorter survival to second-line docetaxel. Breast cancer susceptibility gene 1, MMSET and UBC9 can be useful for customising chemotherapy in gastric cancer patients.