Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML2300

Sigma-Aldrich

ADA-07

≥95% (HPLC)

Synonym(s):

5-(adamantan-1-yl)-3-(hydroxyimino) indolin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20N2O2
CAS Number:
Molecular Weight:
296.36
UNSPSC Code:
12352200
NACRES:
NA.77
Product SML2300 is not currently sold in your country. Contact Technical Service

assay

≥95% (HPLC)

form

powder

color

yellow to brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(/C1=N\O)NC(C1=C2)=CC=C2C34C[C@@H](C5)C[C@@H](C[C@@H]5C4)C3

Biochem/physiol Actions

ADA-07 is a potent and selective T-LAK cell-originated protein kinase (TOPK) inhibitor that binds to ATP-binding pocket. ADA-07 suppresses SUV-induced phosphorylation of ERK1/2, p38, and JNKs and subsequently inhibited AP-1 activity, and reduces SUV-induced skin carcinogenesis.
potent and selective TOPK inhibitor that binds to ATP-binding pocket; reduces SUV-induced skin carcinogenesis

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ge Gao et al.
Molecular cancer therapeutics, 16(9), 1843-1854 (2017-06-29)
Cumulative exposure to solar ultraviolet (SUV) irradiation is regarded as the major etiologic factor in the development of skin cancer. The activation of the MAPK cascades occurs rapidly and is vital in the regulation of SUV-induced cellular responses. The T-LAK

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service