Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0730

Sigma-Aldrich

PCNA-I1

≥98% (HPLC)

Synonym(s):

N-(3-Methylthiophene-2-carboxylic acid)-N′-[(3-hydroxy-2-naphthalenyl)methylene]hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14N2O2S
CAS Number:
Molecular Weight:
310.37
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

PCNA-I1 is a small molecule PCNA inhibitor that selectively binds to PCNA trimers and reduces the association of PCNA with chromatin. PCNA-I1 induces S and G2/M phase cell cycle arrest in tumor cells with greater efficacy than non-transformed cells.
Proliferating cell nuclear antigen-l1 (PCNA) is a member of the scaffold B class. It acts as a linker at the monomer-monomer interface. PCNA-I1 has the ability to block the development of primary cultures of bone marrow mesenchymal stem cells, endothelial cells, lymphocytes, mammary epithelial cells and prostate epithelial cells.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Target validation and structure-activity analysis of a series of novel PCNA inhibitors
Dillehay KL, et al.
Pharmacology Research & Perspectives, 3(2), e00115-e00115 (2015)
Small molecule targeting of PCNA chromatin association inhibits tumor cell growth
Tan Z, et al.
Molecular Pharmacology, mol-112 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service