Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML1946

Sigma-Aldrich

AM-9074

≥98% (HPLC)

Synonym(s):

(R)-1-(4-((5-Methoxypyridin-3-yl)oxy)-5-(tetrahydro-2H-pyran-3-yl)pyridin-2-yl)-3-methylurea, Urea, N-[4-[(5-methoxy-3-pyridinyl)oxy]-5-[(3R)-tetrahydro-2H-pyran-3-yl]-2-pyridinyl]-N′-methyl-

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$92.05
25 MG
$391.00

$92.05

List Price$96.90Save 5%
Web-Only Promotion

Available to ship onMay 02, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
5 MG
$92.05
25 MG
$391.00

About This Item

Empirical Formula (Hill Notation):
C18H22N4O4
CAS Number:
Molecular Weight:
358.39
UNSPSC Code:
12352200
NACRES:
NA.77

$92.05

List Price$96.90Save 5%
Web-Only Promotion

Available to ship onMay 02, 2025Details


Request a Bulk Order
Notify Me

Get notified when this item is ready to ship via email.

Quality Level

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CNC(NC1=CC(OC2=CC(OC)=CN=C2)=C([C@H]3CCCOC3)C=N1)=O

InChI

1S/C18H22N4O4/c1-19-18(23)22-17-7-16(26-14-6-13(24-2)8-20-9-14)15(10-21-17)12-4-3-5-25-11-12/h6-10,12H,3-5,11H2,1-2H3,(H2,19,21,22,23)/t12-/m0/s1

InChI key

APSOPISMEDAYTH-LBPRGKRZSA-N

Biochem/physiol Actions

AM-9074 is a potent activator of glucokinase, which catalyzes the phosphorylation of glucose to glucose-6-phosphate. AM-9074 had an EC50 value of 160 nM with good solubility, oral bioavailability and acceptable clearance in rodents, and was shown to lower plasma glucose levels.
Glucokinase activator

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Todd J Kohn et al.
ACS medicinal chemistry letters, 7(7), 666-670 (2016-07-21)
Two 1-(4-aryl-5-alkyl-pyridin-2-yl)-3-methylurea glucokinase activators were identified with robust in vivo efficacy. These two compounds possessed higher solubilities than the previously identified triaryl compounds (i.e., AM-2394). Structure-activity relationship studies are presented along with relevant pharmacokinetic and in vivo data.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service