Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0967

Sigma-Aldrich

TAE-1

≥98% (HPLC)

Synonym(s):

2,2′,2′-[1,3,5-Triazine-2,4,6-triyltris(oxy-4,1-phenylenecarbonyloxy)]tris[N,N,N-trimethyl-ethanaminium tri-iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H51N6O9 · 3I
CAS Number:
Molecular Weight:
1128.57
UNSPSC Code:
12352200
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

TAE-1 is an inhibitor of amyloid-β fibril formation and aggregation. TAE-1 also inhibits cholinesterases AChE and BuChE.
TAE-1 is an inhibitor of amyloid-β fibril formation and aggregation. TAE-1 is a sym-triazine with dual activity against Alzheimer′s disease. TAE-1 inhibits cholinesterases AChE and BuChE with IC50 values of 0.3 and 3.9 μM. Studies using TAE-1 on differentiated human SH-SY5Y neuronal cells have suggested that it can stimulate synapse formation and promote the differentiation of human neurons, increasing neurite extension.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service