Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0643

Sigma-Aldrich

M617 trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

Galanin(1-13)-Gln14-bradykinin(2-9)amide trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C112H161N29O28 · xC2HF3O2
CAS Number:
Molecular Weight:
2361.65 (free base basis)
UNSPSC Code:
12352202

assay

≥95% (HPLC)

form

powder

color

white to beige

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C112H161N29O28/c1-59(2)44-73(130-97(155)74(45-60(3)4)131-99(157)77(49-66-33-35-68(145)36-34-66)127-90(149)54-121-95(153)62(7)124-104(162)81(57-142)135-102(160)80(51-88(115)147)132-98(156)75(46-61(5)6)134-108(166)93(63(8)144)137-103(161)79(125-89(148)52-113)50-67-53-120-70-27-16-15-26-69(67)70)96(154)123-56-92(151)138-40-18-30-84(138)106(164)129-72(37-38-87(114)146)109(167)141-43-21-32-86(141)111(169)140-42-19-29-83(140)105(163)122-55-91(150)126-76(47-64-22-11-9-12-23-64)100(158)136-82(58-143)110(168)139-41-20-31-85(139)107(165)133-78(48-65-24-13-10-14-25-65)101(159)128-71(94(116)152)28-17-39-119-112(117)118/h9-16,22-27,33-36,53,59-63,71-86,93,120,142-145H,17-21,28-32,37-52,54-58,113H2,1-8H3,(H2,114,146)(H2,115,147)(H2,116,152)(H,121,153)(H,122,163)(H,123,154)(H,124,162)(H,125,148)(H,126,150)(H,127,149)(H,128,159)(H,129,164)(H,130,155)(H,131,157)(H,132,156)(H,133,165)(H,134,166)(H,135,160)(H,136,158)(H,137,161)(H4,117,118,119)/t62-,63+,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,93-/m0/s1

Inchi Key

PAYBRARNTYCYOT-OTTBQUPZSA-N

Amino Acid Sequence

GLY-TRP-THR-LEU-ASN-SER-ALA-GLY-TYR-LEU-LEU-GLY-PRO-GLN-PRO-PRO-GLY-PHE-SER-PRO-PHE-ARG-NH2 trifluoroacetate salt

Biochem/physiol Actions

M617, galanin(1–13)-Gln14-bradykinin(2–9)amide, is a chimeric peptide that acts as a selective agonist for the Galanin GalR1 receptor. M617 shows 25-fold specificity for GalR1with Ki values of 0.23 for GAL1 and 5.71 nM for GAL2.
M617is a Galanin GalR1 selective agonist.

Features and Benefits

This compound is featured on the Galanin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to galanin receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service