Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML0402

Sigma-Aldrich

PD 151746

≥98% (HPLC)

Synonym(s):

3-(5-Fluoro-1H-indol-3-yl)-2-mercapto-2-propenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8FNO2S
CAS Number:
Molecular Weight:
237.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

light orange to dark orange

solubility

DMSO: 15 mg/mL (clear orange solution)

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)\C(S)=C\c1c[nH]c2ccc(F)cc12

InChI

1S/C11H8FNO2S/c12-7-1-2-9-8(4-7)6(5-13-9)3-10(16)11(14)15/h1-5,13,16H,(H,14,15)/b10-3-

InChI key

HWMQHECFXSVZGN-KMKOMSMNSA-N

Biochem/physiol Actions

PD 151746 is a calpain 1 and 2 inhibitor.
PD 151746 is a potent inhibitor of calpains 1 and 2.

Other Notes

air sensitive

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service