Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4300527

Sigma-Aldrich

Anti-RPS6KB1 (Ab-424) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-PS6K antibody produced in rabbit, Anti-S6K antibody produced in rabbit, Anti-S6K1 antibody produced in rabbit, Anti-STK14A antibody produced in rabbit, Anti-ribosomal protein S6 kinase, 70kDa, polypeptide 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$341.00

$341.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$341.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$341.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~70 kDa
~85 kDa

species reactivity

human, mouse, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(P-V-S-P-V)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPS6KB1(6198)

Immunogen

Peptide sequence around aa. 422-426 (P-V-S-P-V), according to the protein RPS6KB1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

RPS6KB1 phosphorylates the Ribosomal Protein-S6. Activation of RPS6KB1 requires a complex, ordered series of conformational changes and phosphorylation reactions. While the role of sequential, multi-site phosphorylation has been extensively detailed, characterization of the priming step required to initiate this cascade has remained elusive. Probably this priming process is dependent on calcium. Calcium-dependent regulation of RPS6KB1 does not specifically target Thr-229 and Thr-389, the key regulatory phosphorylation sites; rather, calcium chelation results in a global inhibition of RPS6KB1 phosphorylation. The initial calcium-dependent process is required to release an inhibitory interaction between the C- and N-termini of RPS6KB1, thus allowing phosphorylation of key domains. The priming event involves formation of a calcium-dependent protein complex that releases the interaction between the N- and C-termini. RPS6KB1 is then accessible for activation by the kinases that target the known regulatory phosphorylation sites .

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service