Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4300503

Sigma-Aldrich

ANTI-TAU (MAPT) Antibody

rabbit polyclonal

Synonym(s):

Anti-DDPAC antibody produced in rabbit, Anti-FLJ31424 antibody produced in rabbit, Anti-FTDP-17 antibody produced in rabbit, Anti-MAPTL antibody produced in rabbit, Anti-microtubule-associated protein tau antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$341.00

$341.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1092


Select a Size

Change View
100 μG
$341.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$341.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1092

Product Name

Anti-MAPT (Ab-212) antibody produced in rabbit, affinity isolated antibody

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~48 kDa
~62 kDa
~78 kDa

species reactivity

human, mouse, rat

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(S-R-T-P-S)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPT(4137)

Immunogen

Peptide sequence around aa. 210-214 (S-R-T-P-S), according to the protein MAPT.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Promotes microtubule assembly and stability, and might be involved in the establishment and maintenance of neuronal polarity. The C-terminus binds axonal microtubules while the N-terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by tau localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton whereas the longer isoforms may preferentially play a role in its stabilization.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service