Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200851

Sigma-Aldrich

Anti-Proteus mirabilis LPS antibody, Mouse monoclonal

clone PMIR174, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

PMIR174, monoclonal

description

Research area: Microbiome

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoblotting: 0.25-0.5 μg/mL using using Proteus mirabilis whole bacteria lysate.
indirect ELISA: 0.25-0.5 μg/mL using Proteus mirabilis LPS for coating.

isotype

IgG2b

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Proteus mirabilis is a Gram negative rod-shaped bacteria, belongs to the Enterobacteriaceae family. Member of the Proteus genus (Proteus spp.) which also includes Proteus mirabilis, Proteus penneri and Proteus hauseri, originally characterize by their ability to swarm on solid surfaces, are widespread in the environment and the gastrointestinal tract of human and animals and known to be an opportunistic pathogens isolated from urine, wounds and other clinical sources. The Proteus spp. bacteria, are distinguished by their reactions for indole production, salicin fermentation and aesculin hydrolysis.1-3 P. vulgaris produces indole which differentiates it from the indole-negative P. mirabilis and P. penneri.1-3 Proteus spp. bacteria may also be found in soil or water habitats where they often regarded as indicators of fecal pollution and a contamination threat for potential water or seafood poisoning.1

Specificity

Monoclonal Anti-Proteus mirabilis LPS specifically recognizes P. mirabilis whole extract and P. mirabilis LPS, the antibody has no cross reactivity with P. vulgaris LPS nor with whole bacterial extracts of P. vulgaris, P. gingivalis, E. coli K-12, P. aeruginosa, S. flexneri, S. enterica, E. faecalis and B. subtilis.

Immunogen

Proteus Mirabilis LPS

Application

The antibody may be used in various immunochemical techniques including Immunoblotting and ELISA. Detection of the P. mirabilis LPS bands by Immunoblotting is specifically inhibited by the immunogen.

Biochem/physiol Actions

In the Proteus spp. group, P. mirabilis is encountered in the community and causes the majority of urinary tract Proteus spp. infections whereas P. vulgaris and P. penneri are less common and mainly associated with nosocomial none urinary infections.2,4P. mirabilis have a number of putative virulence factors including urease, haemolysin, fimbriae structure and iron uptake which has been suggested to contribute to host cell invasion and cytotoxicity, ability to invade uroepithelial cells and agglutinate red blood cells. 4 P. vulgaris, P. mirabilis and P. penneri harbor resistance to ß-lactam antibiotics as it is capable of producing inducible ß-lactamases that hydrolyze primary and extended-spectrum penicillins and cephalosporins.5-7

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service